Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-81744 NB110-81744SS
The SIRT7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SIRT7. This antibody reacts with human, mouse. The SIRT7 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SIRT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.025 ml NB110-81744SS 102125-732 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

100 μl NB110-81744 102125-730 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SIRT7
Antigen Name Sirtuin 7
Reactivity Human, Mouse Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoFluorescence Yes
Antigen Synonyms sirtuin 7, EC 3.5.1.-, homolog) 7, SIR2L7NAD-dependent deacetylase sirtuin-7, MGC126840, S.cerevisiae, sir2-related protein type 7, SIR2-like protein 7, sirtuin type 7, silent mating type information regulation 2, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), MGC126842, homolog 7, sirtuin (silent mating type information regulation 2 sir2-related protein type 7, SIR2-like protein 7, MGC126840, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), SIR2L7NAD-dependent deacetylase sirtuin-7, S.cerevisiae, EC 3.5.1.-, homolog) 7, sirtuin (silent mating type information regulation 2, MGC126842, homolog 7, sirtuin type 7, silent mating type information regulation 2, sirtuin 7
Gene ID 51547
Concentration 1 mg/ml
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 100-200). [Swiss-Prot# Q9NRC8]
Purification Immunogen affinity purified
Storage Buffer Tris-Citrate/Phosphate (pH 7.0 - 8.0)
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty