Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB100-2524R
102107-664EA 614.15 USD
102107-664
Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The SIRT6 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to SIRT6. This antibody reacts with human. The SIRT6 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR