Anti-SIRT3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31029
102164-022EA 635.51 USD
102164-022
Anti-SIRT3 Rabbit Polyclonal Antibody
Antibodies
The Sirtuin 3 / SIRT3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Sirtuin 3 / SIRT3. This antibody reacts with human. The Sirtuin 3 / SIRT3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR