Anti-SIRT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

NBP1-49540H
102179-176EA 490.77 USD
102179-176
Anti-SIRT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
The Sirtuin 1 / SIRT1 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to Sirtuin 1 / SIRT1. This antibody reacts with human. The Sirtuin 1 / SIRT1 Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR