Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT1 Rabbit Polyclonal Antibody (DyLight 650)

Supplier: Novus Biologicals
NBP1-49540C
102179-172
Anti-SIRT1 Rabbit Polyclonal Antibody (DyLight 650)
The Sirtuin 1 / SIRT1 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to Sirtuin 1 / SIRT1. This antibody reacts with human. The Sirtuin 1 / SIRT1 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty