Anti-SIPA1L2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-72138
102201-976EA 399 USD
102201-976
Anti-SIPA1L2 Rabbit Polyclonal Antibody
Antibodies
The SIPA1L2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SIPA1L2. This antibody reacts with human, mouse, rat. The SIPA1L2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: SIPA1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR