Anti-SIK1/Snf1lk Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-82417
102216-894EA 564.74 USD
102216-894
Anti-SIK1/Snf1lk Rabbit Polyclonal Antibody
Antibodies
The SIK1 / Snf1lk Antibody from Novus Biologicals is a rabbit polyclonal antibody to SIK1 / Snf1lk. This antibody reacts with rat. The SIK1 / Snf1lk Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SIK1/Snf1lk
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR