Anti-Signal peptide peptidase-like 2B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59510
102192-712EA 564.74 USD
102192-712
Anti-Signal peptide peptidase-like 2B Rabbit Polyclonal Antibody
Antibodies
The Signal peptide peptidase-like 2B Antibody from Novus Biologicals is a rabbit polyclonal antibody to Signal peptide peptidase-like 2B. This antibody reacts with human. The Signal peptide peptidase-like 2B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Signal peptide peptidase-like 2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR