Anti-Siglec-11 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77025
102210-300EA 564.74 USD
102210-300
Anti-Siglec-11 Rabbit Polyclonal Antibody
Antibodies
The Siglec-11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Siglec-11. This antibody reacts with human. The Siglec-11 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Siglec-11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR