Anti-Siglec-7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59237
102192-180EA 564.74 USD
102192-180
Anti-Siglec-7 Rabbit Polyclonal Antibody
Antibodies
The Siglec-7 / CD328 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Siglec-7 / CD328. This antibody reacts with human. The Siglec-7 / CD328 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Siglec-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR