Anti-SIDT2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91551
102217-640EA 564.74 USD
102217-640
Anti-SIDT2 Rabbit Polyclonal Antibody
Antibodies
The SIDT2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SIDT2. This antibody reacts with human. The SIDT2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SIDT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR