Anti-ST3GAL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-62540
102195-706EA 564.74 USD
102195-706
Anti-ST3GAL1 Rabbit Polyclonal Antibody
Antibodies
The ST3 beta-Gal alpha-2,3-Sialyltransferase 1 / ST3GAL1 / SIAT4A Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST3 beta-Gal alpha-2,3-Sialyltransferase 1 / ST3GAL1 / SIAT4A. This antibody reacts with human. The ST3 beta-Gal alpha-2,3-Sialyltransferase 1 / ST3GAL1 / SIAT4A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ST3GAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR