Anti-SHD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78137
102212-528EA 805.35 USD
102212-528
Anti-SHD Rabbit Polyclonal Antibody
Antibodies
The SHD Antibody from Novus Biologicals is a rabbit polyclonal antibody to SHD. This antibody reacts with drosophila. The SHD Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SHD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR