Anti-SH3BGR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80550
102216-746EA 564.74 USD
102216-746
Anti-SH3BGR Rabbit Polyclonal Antibody
Antibodies
The SH3BGR Antibody from Novus Biologicals is a rabbit polyclonal antibody to SH3BGR. This antibody reacts with human. The SH3BGR Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SH3BGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR