Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-45853
102175-274EA 706.28 USD
102175-274
Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody
Antibodies
The SGLT2 / SLC5A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SGLT2 / SLC5A2. This antibody reacts with human, mouse, rat. The SGLT2 / SLC5A2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SGLT2/SLC5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR