Anti-SGLT1/SLC5A1 Rabbit Polyclonal Antibody

NBP1-46008
102175-382EA 466.15 USD
102175-382
Anti-SGLT1/SLC5A1 Rabbit Polyclonal Antibody
The SGLT1 / SLC5A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SGLT1 / SLC5A1. This antibody reacts with human. The SGLT1 / SLC5A1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SGLT1/SLC5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR