Anti-SGK3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NLS7240
102253-464EA 329 USD
102253-464
Anti-SGK3 Rabbit Polyclonal Antibody
Antibodies
The SGK3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SGK3. This antibody reacts with human, mouse, rat, bat, canine, hamster. The SGK3 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SGK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bat, Canine, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR