Anti-RBM27 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-60426
102110-942EA 635.51 USD
102110-942
Anti-RBM27 Rabbit Polyclonal Antibody
Antibodies
The RBM27 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RBM27. This antibody reacts with human, mouse. The RBM27 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500

Type: Primary
Antigen: RBM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR