Anti-RAI14 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91459
102217-456EA 564.74 USD
102217-456
Anti-RAI14 Rabbit Polyclonal Antibody
Antibodies
The RAI14 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RAI14. This antibody reacts with human. The RAI14 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: RAI14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR