Anti-RAB6C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74265
102205-270EA 564.74 USD
102205-270
Anti-RAB6C Rabbit Polyclonal Antibody
Antibodies
The RAB6C Antibody from Novus Biologicals is a rabbit polyclonal antibody to RAB6C. This antibody reacts with human. The RAB6C Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: RAB6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR