Anti-Proteasome subunit beta type 4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54586
102184-140EA 564.74 USD
102184-140
Anti-Proteasome subunit beta type 4 Rabbit Polyclonal Antibody
Antibodies
The Proteasome subunit beta type 4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Proteasome subunit beta type 4. This antibody reacts with human. The Proteasome subunit beta type 4 Antibody has been validated for the following applications: Western Blot.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: Proteasome subunit beta type 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR