Anti-Proteasome 20S LMP7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33540
102167-142EA 635.51 USD
102167-142
Anti-Proteasome 20S LMP7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Proteasome 20S LMP7 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: Proteasome 20S LMP7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR