Anti-Oxytocin Rabbit Polyclonal Antibody

NB600-667
102134-426EA 423.08 USD
102134-426
Anti-Oxytocin Rabbit Polyclonal Antibody
The Oxytocin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Oxytocin. This antibody reacts with rat. The Oxytocin Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Oxytocin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR