Anti-OXSR1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-77321
102116-908EA 635.51 USD
102116-908
Anti-OXSR1 Rabbit Polyclonal Antibody
Antibodies
The OXSR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OXSR1. This antibody reacts with human. The OXSR1 Antibody has been validated for the following applications: Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: OXSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR