Anti-OTX3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80255
102216-158EA 564.74 USD
102216-158
Anti-OTX3 Rabbit Polyclonal Antibody
Antibodies
The OTX3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OTX3. This antibody reacts with human, mouse. The OTX3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OTX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR