Anti-Otx1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-12545
102251-954EA 660 USD
102251-954
Anti-Otx1 Rabbit Polyclonal Antibody
Antibodies
The Otx1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Otx1. This antibody reacts with human. The Otx1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Otx1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR