Anti-OCN Rabbit Polyclonal Antibody

NB600-1456
102133-356EA 423.08 USD
102133-356
Anti-OCN Rabbit Polyclonal Antibody
The Osteocalcin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Osteocalcin. This antibody reacts with human. The Osteocalcin Antibody has been validated for the following applications: ELISA, Radioimmunoassay.

Type: Primary
Antigen: OCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR