Anti-OSBPL6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31456
102164-664EA 635.51 USD
102164-664
Anti-OSBPL6 Rabbit Polyclonal Antibody
Antibodies
The OSBPL6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OSBPL6. This antibody reacts with human. The OSBPL6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OSBPL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR