Anti-OSBPL3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-55151
102185-250EA 635.51 USD
102185-250
Anti-OSBPL3 Rabbit Polyclonal Antibody
Antibodies
The OSBPL3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OSBPL3. This antibody reacts with human, mouse. The OSBPL3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: OSBPL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR