Anti-OS9 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NB100-519G
102109-468EA 614.15 USD
102109-468
Anti-OS9 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The OS9 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to OS9. This antibody reacts with human, mouse. The OS9 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250

Type: Primary
Antigen: OS9
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR