Anti-Orc2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46175
102175-702EA 635.51 USD
102175-702
Anti-Orc2 Rabbit Polyclonal Antibody
Antibodies
The Orc2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Orc2. This antibody reacts with human. The Orc2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.

Type: Primary
Antigen: Orc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR