Anti-OR13C9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79986
102215-620EA 564.74 USD
102215-620
Anti-OR13C9 Rabbit Polyclonal Antibody
Antibodies
The OR13C9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OR13C9. This antibody reacts with human. The OR13C9 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OR13C9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR