Anti-OPA1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB110-55290B
102122-526EA 614.15 USD
102122-526
Anti-OPA1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
The OPA1 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to OPA1. This antibody reacts with human, mouse, rat, porcine, chicken. The OPA1 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OPA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR