Anti-OMA1 Rabbit Polyclonal Antibody

NBP1-56970
102188-854EA 493.85 USD
102188-854
Anti-OMA1 Rabbit Polyclonal Antibody
The OMA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OMA1. This antibody reacts with human, fish. The OMA1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: OMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR