Anti-Olig1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-28666
102162-506EA 491.08 USD
102162-506
Anti-Olig1 Rabbit Polyclonal Antibody
Antibodies
The Olig1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Olig1. This antibody reacts with human, mouse, rat. The Olig1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000

Type: Primary
Antigen: Olig1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR