Anti-OIP106 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-56761
102188-442EA 564.74 USD
102188-442
Anti-OIP106 Rabbit Polyclonal Antibody
Antibodies
The OIP106 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OIP106. This antibody reacts with human. The OIP106 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: OIP106
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR