Anti-O-GlcNAc Transferase p110 subunit Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32791
102166-390EA 635.51 USD
102166-390
Anti-O-GlcNAc Transferase p110 subunit Rabbit Polyclonal Antibody
Antibodies
The O-GlcNAc Transferase p110 subunit Antibody from Novus Biologicals is a rabbit polyclonal antibody to O-GlcNAc Transferase p110 subunit. This antibody reacts with human, mouse, rat. The O-GlcNAc Transferase p110 subunit Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: O-GlcNAc Transferase p110 subunit
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR