Anti-OCT4 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB100-2379R
102107-242EA 614.15 USD
102107-242
Anti-OCT4 Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The OCT4 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to 43012. This antibody reacts with human, mouse, rat, primate. The OCT4 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: OCT4
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR