Anti-NUP50 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-93324
102120-774EA 635.51 USD
102120-774
Anti-NUP50 Rabbit Polyclonal Antibody
Antibodies
The NUP50 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NUP50. This antibody reacts with human, mouse. The NUP50 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: NUP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR