Anti-NUDT21 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57540
102189-964EA 564.74 USD
102189-964
Anti-NUDT21 Rabbit Polyclonal Antibody
Antibodies
The NUDT21 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NUDT21. This antibody reacts with human. The NUDT21 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: NUDT21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR