Anti-NUP62 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32177
102165-596EA 635.51 USD
102165-596
Anti-NUP62 Rabbit Polyclonal Antibody
Antibodies
The NUP62 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NUP62. This antibody reacts with human, mouse, rat. The NUP62 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: NUP62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR