Anti-NPM1 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB110-61646R
102124-526EA 626.46 USD
102124-526
Anti-NPM1 Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The NPM1 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to NPM1. This antibody reacts with human. The NPM1 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 - 1:100

Type: Primary
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR