Anti-NTAL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-44948
102174-410EA 309 USD
102174-410
Anti-NTAL Rabbit Polyclonal Antibody
Antibodies
The NTAL Antibody from Novus Biologicals is a rabbit polyclonal antibody to NTAL. This antibody reacts with human, mouse (negative). The NTAL Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: NTAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR