Anti-NTAL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31177
102164-252EA 635.51 USD
102164-252
Anti-NTAL Rabbit Polyclonal Antibody
Antibodies
The NTAL Antibody from Novus Biologicals is a rabbit polyclonal antibody to NTAL. This antibody reacts with human. The NTAL Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: NTAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR