Anti-NT5C1B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-56911
102188-736EA 564.74 USD
102188-736
Anti-NT5C1B Rabbit Polyclonal Antibody
Antibodies
The NT5C1B Antibody from Novus Biologicals is a rabbit polyclonal antibody to NT5C1B. This antibody reacts with human. The NT5C1B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: NT5C1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR