Anti-MUC1 Rabbit Polyclonal Antibody

NB300-708
102131-004EA 635.38 USD
102131-004
Anti-MUC1 Rabbit Polyclonal Antibody
Rabbit Polyclonal MUC1 Antibody. Tested Applications: Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR