Anti-Mu Opioid Receptor Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-79878
102125-420EA 491.08 USD
102125-420
Anti-Mu Opioid Receptor Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Mu Opioid Receptor Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: Mu Opioid Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR