Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TOR/mTOR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-02454
The TOR / mTOR Antibody from Novus Biologicals is a rabbit polyclonal antibody to TOR / mTOR. This antibody reacts with human, mouse, rat. The TOR / mTOR Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 2.5 μg/ml

Type: Primary
Antigen: TOR/mTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa2440-2457
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg NBP1-02454 102155-374 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TOR/mTOR
Antigen Name Mechanistic Target of Rapamycin
Reactivity Rat, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms RAPT1FKBP-rapamycin associated protein, FK506-binding protein 12-rapamycin complex-associated protein 1, EC 2.7.11.1, serine/threonine-protein kinase mTOR, rapamycin associated protein FRAP2, mechanistic target of rapamycin (serine/threonine kinase), FK506 binding protein 12-rapamycin associated protein 1, mTOR, FKBP12-rapamycin complex-associated protein, Rapamycin target protein 1, rapamycin and FKBP12 target 1, Mammalian target of rapamycin, FRAP2, FRAP1FKBP12-rapamycin complex-associated protein 1, FLJ44809, Mechanistic target of rapamycin, RAFT1, FK506 binding protein 12-rapamycin associated protein 2, FRAP
Gene ID 2475
Epitope aa2440-2457
Concentration 0.9 mg/ml
Immunogen Synthetic peptide from human mTOR.
Purification Immunogen affinity purified
Storage Buffer 20mM Potassium Phosphate (pH 7.2) and 0.15M NaCl
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping