Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TOR/mTOR Rabbit Polyclonal Antibody (Agarose Immobilized)

Supplier: Novus Biologicals
The TOR / mTOR Antibody [Agarose Immobilized] from Novus Biologicals is a rabbit polyclonal antibody to TOR / mTOR. This antibody reacts with human, mouse, rat. The TOR / mTOR Antibody [Agarose Immobilized] has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: TOR/mTOR
Clonality: Polyclonal
Conjugation: Agarose Immobilized
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB600-425 102133-950 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol TOR/mTOR
Antigen Name Mechanistic Target of Rapamycin
Reactivity Human, Mouse, Rat
Conjugation Agarose Immobilized
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoPrecipitation Yes
Antigen Synonyms FKBP12-rapamycin complex-associated protein, Mammalian target of rapamycin, Rapamycin target protein 1, FRAP2, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FK506-binding protein 12-rapamycin complex-associated protein 1, Mechanistic target of rapamycin, EC, serine/threonine-protein kinase mTOR, rapamycin associated protein FRAP2, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, rapamycin and FKBP12 target 1, FLJ44809, RAPT1FKBP-rapamycin associated protein, RAFT1, mTOR, mechanistic target of rapamycin (serine/threonine kinase)
Gene ID 2475
Concentration 0.1 mg/ml
Immunogen The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Mammalian Target of Rapamycin using the numbering given in entry NP_004949.1 (GeneID 2475).
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty