Anti-MTMR4 Rabbit Polyclonal Antibody

NBP1-32402
102165-872EA 493.85 USD
102165-872
Anti-MTMR4 Rabbit Polyclonal Antibody
Rabbit Polyclonal MTMR4 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: MTMR4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR